Skip to Content
MilliporeSigma
All Photos(1)

Documents

W336300

Sigma-Aldrich

6-Methyl-3,5-heptadien-2-one

≥96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O
CAS Number:
Molecular Weight:
124.18
FEMA Number:
3363
EC Number:
Council of Europe no.:
11143
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.099

grade

Halal

description

predominantly trans

Assay

≥96%

form

liquid

refractive index

n20/D 1.5360 (lit.)

bp

168 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\C=C\C(C)=O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.1 °F - closed cup

Flash Point(C)

80.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service