Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W336009

Sigma-Aldrich

3-Methyl-2-cyclohexenone

≥98%, stabilized, FG

Synonym(s):

nutty cyclohexenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H7(=O)
CAS Number:
Molecular Weight:
110.15
FEMA Number:
3360
Beilstein:
1560601
EC Number:
Council of Europe no.:
11134
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.098
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

contains

α-Tocopherol, synthetic as stabilizer

refractive index

n20/D 1.494 (lit.)

bp

199-200 °C (lit.)

density

0.971 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; nutty; sweet

SMILES string

CC1=CC(=O)CCC1

InChI

1S/C7H10O/c1-6-3-2-4-7(8)5-6/h5H,2-4H2,1H3

InChI key

IITQJMYAYSNIMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoichiro Horita et al.
Chembiochem : a European journal of chemical biology, 16(3), 440-445 (2015-02-03)
(4R,6R)-Actinol can be stereo-selectively synthesized from ketoisophorone by a two-step conversion using a mixture of two enzymes: Candida macedoniensis old yellow enzyme (CmOYE) and Corynebacterium aquaticum (6R)-levodione reductase. However, (4S)-phorenol, an intermediate, accumulates because of the limited substrate range of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service