Skip to Content
MilliporeSigma
All Photos(2)

Documents

W335900

Sigma-Aldrich

2-Methylbutyl 2-methylbutyrate

natural, ≥98%, FG

Synonym(s):

DL-2-Methylbutyric acid 2-methylbutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2CH2CH(CH3)C2H5
CAS Number:
Molecular Weight:
172.26
FEMA Number:
3359
EC Number:
Council of Europe no.:
10773
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.516
NACRES:
NA.21

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.411-1.417 (lit.)

bp

70-72 °C/11 mmHg (lit.)

density

0.855 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green; waxy; fruity; sweet

SMILES string

CCC(C)COC(=O)C(C)CC

InChI

1S/C10H20O2/c1-5-8(3)7-12-10(11)9(4)6-2/h8-9H,5-7H2,1-4H3

InChI key

PVYFCGRBIREQLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F

Flash Point(C)

67 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service