Skip to Content
MilliporeSigma
All Photos(1)

Documents

W335118

Sigma-Aldrich

3-Hexanol

≥97%

Synonym(s):

Ethyl propyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH2CH3
CAS Number:
Molecular Weight:
102.17
FEMA Number:
3351
Beilstein:
1718964
EC Number:
Council of Europe no.:
4123
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.089
NACRES:
NA.21

biological source

synthetic

Assay

≥97%

refractive index

n20/D 1.415 (lit.)

bp

134-135 °C (lit.)

density

0.820 g/mL at 20 °C
0.819 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

alcohol; medicinal; ethereal

SMILES string

CCCC(O)CC

InChI

1S/C6H14O/c1-3-5-6(7)4-2/h6-7H,3-5H2,1-2H3

InChI key

ZOCHHNOQQHDWHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Hexanol is an aliphatic alcohol that is reported to occur in freesia oil, pandan leaves and in tomato and dried products.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile compounds in tomato-based dried products
Narain N, et al.
Drying Technology, 28(2), 232-239 (2010)
The volatile constituents of freesia flower (Freesia hybrida Hort.).
Harada K & Mihara S.
Agricultural and Biological Chemistry, 48(11), 2843-2845 (1984)
Volatile composition of pandan leaves (Pandanus amaryllifolius).
Jiang J.
Flavor chemistry of ethnic foods, 105-109 (1999)
Belén Cotes et al.
Oecologia, 179(1), 151-162 (2015-05-07)
To locate and evaluate host patches before oviposition, parasitoids of herbivorous insects utilize plant volatiles and host-derived cues, but also evaluate predator-derived infochemicals to reduce predation risks. When foraging in host habitats infested with entomopathogenic fungi that can infect both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service