Skip to Content
MilliporeSigma
All Photos(1)

Documents

W330302

Sigma-Aldrich

2-Methyl-1-butanethiol

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH2SH
CAS Number:
Molecular Weight:
104.21
FEMA Number:
3303
EC Number:
Council of Europe no.:
11509
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.048
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

vapor pressure

41.4 mmHg ( 37.7 °C)

Assay

≥95%

refractive index

n20/D 1.447 (lit.)

bp

116-117 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

SMILES string

CCC(C)CS

InChI

1S/C5H12S/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3

InChI key

WGQKBCSACFQGQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service