Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W328618

Sigma-Aldrich

Tripropionin

≥97.1%, FCC, FG

Synonym(s):

Glyceryl tripropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5CO2CH2)2CH(O2CC2H5)
CAS Number:
Molecular Weight:
260.28
FEMA Number:
3286
EC Number:
Council of Europe no.:
10657
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.263
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC

vapor density

8.95 (vs air)

Assay

≥97.1%

autoignition temp.

790 °F

expl. lim.

0.8 %, 186 °F

refractive index

n20/D 1.434 (lit.)

bp

175-176 °C/20 mmHg (lit.)

density

1.08 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

oily; chemical

SMILES string

CCC(=O)OCC(COC(=O)CC)OC(=O)CC

InChI

1S/C12H20O6/c1-4-10(13)16-7-9(18-12(15)6-3)8-17-11(14)5-2/h9H,4-8H2,1-3H3

InChI key

YZWRNSARCRTXDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Lombardo et al.
Biochimica et biophysica acta, 659(2), 401-410 (1981-06-15)
Human pancreatic carboxylic ester hydrolase (EC 3.1.1.1), usually characterized by its activity on water-soluble substrates, is shown to catalyze reactions taking place at a lipid/water interface. The inhibition of tributyrin hydrolysis by 1-alcohols follows the pattern of a Langmuir adsorption
Slim Cherif et al.
Lipids in health and disease, 10, 221-221 (2011-11-30)
Bacterial lipases received much attention for their substrate specificity and their ability to function in extreme environments (pH, temperature...). Many staphylococci produced lipases which were released into the culture medium. Reports of thermostable lipases from Staphylococcus sp. and active in
Riadh Ben Salah et al.
FEMS microbiology letters, 260(2), 241-248 (2006-07-18)
A novel strain of Rhizopus oryzae WPG secretes a noninduced lipase (ROLw) in the culture medium; purified ROLw is a protein of 29 kDa, the 45 N-terminal amino acid residues were sequenced, this sequence is very homologous to Rhizopus delemar
Ahmed Fendri et al.
Archives of biochemistry and biophysics, 451(2), 149-159 (2006-06-20)
Chicken pancreatic lipase (CPL) was purified from delipidated pancreas. Pure CPL was obtained after ammonium sulphate fractionation, then DEAE-cellulose, Sephacryl S-200 gel filtration, and FPLC Mono-Q Sepharose columns. The pure lipase is a glycosylated monomer having a molecular mass of
Henri Chahinian et al.
Lipids, 37(7), 653-662 (2002-09-10)
The better to characterize enzymes hydrolyzing carboxyl ester bonds (carboxyl ester hydrolases), we have compared the kinetic behavior of various lipases and esterases against solutions and emulsions of vinyl esters and TAG. Short-chain vinyl esters are hydrolyzed at comparable rates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service