Skip to Content
MilliporeSigma
All Photos(1)

Documents

W328405

Sigma-Aldrich

1-Furfurylpyrrole

≥98%, FG

Synonym(s):

1-(2-Furanylmethyl)-1H-pyrrole, N-Furfuryl pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
FEMA Number:
3284
EC Number:
Council of Europe no.:
2317
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.134
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

vapor density

>1 (vs air)

Assay

≥98%

refractive index

n20/D 1.531 (lit.)

bp

76-78 °C/1 mmHg (lit.)

density

1.081 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; vegetable; earthy

SMILES string

C(c1ccco1)n2cccc2

InChI

1S/C9H9NO/c1-2-6-10(5-1)8-9-4-3-7-11-9/h1-7H,8H2

InChI key

BTBFUBUCCJKJOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Furfurylpyrrole has been identified as a potential contributor of flavor and aroma to popcorn. It is also found in roasted almonds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavor components of roasted almond.
Takei Y & Yamanishi T.
Agricultural and Biological Chemistry, 38(12), 2329-2336 (1974)
Popcorn flavor: identification of volatile compounds.
Walradt JP, et al.
Journal of Agricultural and Food Chemistry, 18(5), 926-928 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service