Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W319519

Sigma-Aldrich

2-Methyl-2-pentenoic acid

natural (US), 97.0%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
FEMA Number:
3195
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

grade

FG
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

97.0%

refractive index

n20/D 1.460

density

0.982 g/mL

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

pungent; acidic; fruity; sweet; woody

SMILES string

CC/C=C(C)/C(O)=O

InChI

1S/C6H10O2/c1-3-4-5(2)6(7)8/h4H,3H2,1-2H3,(H,7,8)/b5-4+

InChI key

JJYWRQLLQAKNAD-SNAWJCMRSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service