Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W317215

Sigma-Aldrich

Hexyl isobutyrate

natural (US), ≥97%, FG

Synonym(s):

Hexyl-2-methylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2(CH2)5CH3
CAS Number:
Molecular Weight:
172.26
FEMA Number:
3172
EC Number:
Council of Europe no.:
646
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.478
grade:
FG
Halal
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.413 (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green; fruity; pear; sweet

SMILES string

CCCCCCOC(=O)C(C)C

InChI

1S/C10H20O2/c1-4-5-6-7-8-12-10(11)9(2)3/h9H,4-8H2,1-3H3

InChI key

CYHBDKTZDLSRMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhilin Zhang et al.
Journal of insect science (Online), 14 (2014-12-21)
The morphological structure of the metathoracic scent glands (MTGs) in Adelphocoris suturalis was observed by utilizing scanning electron microscope (SEM). Also, the secretions of MTGs in male and female were analyzed by using gas chromatography-mass spectrometry (GC-MS). The result showed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service