Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W307505

Sigma-Aldrich

p-Tolyl isobutyrate

≥97%, FCC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2C6H4CH3
CAS Number:
Molecular Weight:
178.23
FEMA Number:
3075
EC Number:
Council of Europe no.:
304
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.429
NACRES:
NA.21

biological source

synthetic

Quality Level

reg. compliance

FCC

Assay

≥97%

refractive index

n20/D 1.487 (lit.)

bp

237 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

green; lily; fruity; animal; floral

SMILES string

CC(C)C(=O)Oc1ccc(C)cc1

InChI

1S/C11H14O2/c1-8(2)11(12)13-10-6-4-9(3)5-7-10/h4-8H,1-3H3

InChI key

UPPSFGGDKACIKP-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service