Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W305707

Sigma-Aldrich

Tetrahydrofurfuryl butyrate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O3
CAS Number:
Molecular Weight:
172.22
FEMA Number:
3057
EC Number:
Council of Europe no.:
2081
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.048

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.44 (lit.)

bp

225-227 °C/759 mmHg (lit.)

density

1.012 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cheese; pineapple

SMILES string

CCCC(=O)OCC1CCCO1

InChI

1S/C9H16O3/c1-2-4-9(10)12-7-8-5-3-6-11-8/h8H,2-7H2,1H3

InChI key

DPZVDLFOAZNCBU-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service