Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W304514

Sigma-Aldrich

Terpineol

natural

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3045
EC Number:
Council of Europe no.:
62c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.014

grade

Kosher
natural

Quality Level

refractive index

n20/D 1.482-1.485

bp

213-218 °C (lit.)

density

0.934 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

lilac

SMILES string

CC(=C)C1CCC(C)(O)CC1.CC2=CCC(CC2)C(C)(C)O

InChI

1S/2C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;9,11H,1,4-7H2,2-3H3

InChI key

MGABGZGUMNDCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng-Chang Wang et al.
Sensors (Basel, Switzerland), 15(6), 14286-14297 (2015-06-20)
Highly sensitive H2 gas sensors were prepared using pure and Pt-loaded SnO2 nanoparticles. Thick film sensors (~35 μm) were fabricated that showed a highly porous interconnected structure made of high density small grained nanoparticles. Using Pt as catalyst improved sensor
Khaled Sebei et al.
Biological research, 48, 7-7 (2015-02-06)
In this paper, we have studied the essential oils chemical composition of the leaves of seven Eucalyptus species developed in Tunisia. Eucalyptus leaves were picked from trees growing in different arboretums in Tunisia. Choucha and Mrifeg arboretums located in Sedjnene
Yi-Fei Sun et al.
Nanoscale, 7(25), 11173-11181 (2015-06-11)
To date, most investigations of Ni-Fe bimetallic catalysts for solid oxide fuel cells (SOFCs) have focused on materials with micro-scale particle sizes, which severely restrict their catalytic activity. In this study, we fabricated a Ni- and/or Fe-doped A-site-deficient LaSrCrO3 perovskite
Dae-Yong Son et al.
Physical chemistry chemical physics : PCCP, 17(23), 15193-15200 (2015-05-21)
To understand the role of the dye/oxide interface, a model system using a nanocrystalline SnO2 and 3-hexyl thiophene based MK-2 dye is proposed. A thin interfacial TiO2 blocking layer (IBL) is introduced in between SnO2 and MK-2 and its effects
Kuhu Sarkar et al.
ChemSusChem, 8(16), 2696-2704 (2015-06-25)
Diblock copolymers have been used in sol-gel synthesis to successfully tailor the nanoscale morphology of thin ZnO films. As the fabrication of several-micron-thick mesoporous films such as those required in dye-sensitized solar cells (DSSCs) was difficult with this approach, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service