Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W304000

Sigma-Aldrich

Tagete oil

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3040
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Tagetes glandulifera

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012

form

liquid

optical activity

[α]20/D +8.4°, neat

composition

Citronellol, 3%
Eugenol, 0.2%
Limonene, 7.2%
Linalool, 0.5%

refractive index

n20/D 1.481

density

0.886 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; herbaceous; rancid

Analysis Note

Fragrance: Provides fruity, herbaceous, and tobacco like notes

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Asp. Tox. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service