Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W300306

Sigma-Aldrich

Sage oil

Spanish, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3003
MDL number:
UNSPSC Code:
12164502

biological source

Salvia officinalis L.

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

optical activity

[α]20/D +9.0°, neat

origin

Spain origin

refractive index

n20/D 1.468 (lit.)

bp

181 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 - 129.2 °F

Flash Point(C)

47 - 54 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service