Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W298107

Sigma-Aldrich

Rhodinyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
2981
EC Number:
Council of Europe no.:
223c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.033

biological source

synthetic

refractive index

n20/D 1.4540 (lit.)

bp

237 °C (lit.)

density

0.896 g/mL at 25 °C (lit.)

Organoleptic

berry; honey; floral

SMILES string

CC(CCCC(C)=C)CCOC(C)=O

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service