Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W298018

Sigma-Aldrich

Rhodinol

mixture of L-citronellol and geraniol, FCC

Synonym(s):

L-Citronellol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
FEMA Number:
2980
EC Number:
Council of Europe no.:
76
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.027
NACRES:
NA.21
Organoleptic:
floral; waxy; rose
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

optical activity

[α]22/D −6°, neat

refractive index

n20/D 1.469 (lit.)

bp

68-70 °C/1.8 mmHg (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; waxy; rose

SMILES string

C[C@H](CCO)CCCC(C)=C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3

InChI key

QMVPMAAFGQKVCJ-UHFFFAOYSA-N

General description

Natural occurrence: Geranium and citronella oils.

Biochem/physiol Actions

Taste at 0.5 to 10 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 25, 74-74 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service