Skip to Content
MilliporeSigma
All Photos(1)

Documents

W296228

Sigma-Aldrich

(−)-Isopulegol

FCC, ≥95%

Synonym(s):

(1R,2S,5R)-2-Isopropenyl-5-methylcyclohexanol, (1R,3R,4S)-p-Menth-8-en-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2962
Beilstein:
1906573
EC Number:
Council of Europe no.:
2033
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.067

reg. compliance

FCC

Assay

≥95%

optical activity

[α]22/D −7 to 0°, neat

refractive index

n20/D 1.471 (lit.)

bp

212 °C (lit.)

density

0.912 g/mL at 25 °C (lit.)

SMILES string

C[C@@H]1CC[C@H]([C@H](O)C1)C(C)=C

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h8-11H,1,4-6H2,2-3H3/t8-,9+,10-/m1/s1

InChI key

ZYTMANIQRDEHIO-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Olagnier et al.
Bioorganic & medicinal chemistry letters, 17(22), 6075-6078 (2007-10-02)
Pure natural monoterpenes were evaluated in vitro for their antiplasmodial activities against Plasmodium falciparum. Chemically modified terpenes were also tested to see whether the introduction of an alkyne, a cyclopropane, a diene, or a cyclopentenone moiety had an influence on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service