Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W294918

Sigma-Aldrich

Propyl hexanoate

≥98%, FG

Synonym(s):

n-Propyl hexanoate, Caproic acid propyl ester, Propyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2CH2CH2CH3
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2949
EC Number:
Council of Europe no.:
311
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.061
NACRES:
NA.21

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.412 (lit.)

bp

187 °C (lit.)

mp

−69 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; tropical; fruity; pineapple; sweet

SMILES string

CCCCCC(=O)OCCC

InChI

1S/C9H18O2/c1-3-5-6-7-9(10)11-8-4-2/h3-8H2,1-2H3

InChI key

HTUIWRWYYVBCFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service