Skip to Content
MilliporeSigma
All Photos(1)

Documents

W294802

Sigma-Aldrich

Propyl heptanoate

≥98%

Synonym(s):

n-Propyl heptylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CO2CH2CH2CH3
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2948
EC Number:
Council of Europe no.:
367
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.095

Assay

≥98%

refractive index

n20/D 1.417 (lit.)

bp

208 °C (lit.)

mp

−64 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CCCCCCC(=O)OCCC


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service