Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W291609

Sigma-Aldrich

Polysorbate 60

Synonym(s):

TWEEN® 60, Polyethylene glycol sorbitan monostearate, Polyoxyethylene sorbitan monostearate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2916
MDL number:
UNSPSC Code:
12164502

reg. compliance

FDA 21 CFR 172.515
FDA 21 CFR 172.836
FDA 21 CFR 173.340
FDA 21 CFR 175.105

Quality Level

description

derived from GMO corn

refractive index

n20/D 1.474

density

1.044 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

corn

Organoleptic

alcohol

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polysorbate 60 is a hydrophilic nonionic surfactant generally used as an emulsifier, dispersing agent and solubilizer.

Application

Polysorbate 60 has been used in a study to assess its potential as a generally recognized as safe (GRAS) plasticizer in imparting plasticity to carnauba wax.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Food, Drug, and Cosmetic Excipients, 295-296 (1992)
Plasticisation of carnauba wax with generally recognised as safe (GRAS) additives.
Zhang Y, et al.
Polymer, 86, 208-219 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service