Skip to Content
MilliporeSigma
All Photos(1)

Documents

W291307

Sigma-Aldrich

Piperonyl isobutyrate

≥98%, FG

Synonym(s):

Heliotropyl isobutyrate, Isobutyric acid 3,4-methylenedioxybenzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24
FEMA Number:
2913
EC Number:
Council of Europe no.:
305
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.430
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.511 (lit.)

bp

91-92 °C/0.005 mmHg (lit.)

density

1.154 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; jam; fruity

SMILES string

CC(C)C(=O)OCc1ccc2OCOc2c1

InChI

1S/C12H14O4/c1-8(2)12(13)14-6-9-3-4-10-11(5-9)16-7-15-10/h3-5,8H,6-7H2,1-2H3

InChI key

RQULTIASPCVEFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Wright et al.
Xenobiotica; the fate of foreign compounds in biological systems, 10(4), 265-270 (1980-04-01)
1. The metabolism of piperonylidene acetone, and the food additives piperonyl acetone, piperonyl acetate and piperonyl isobutyrate were studied in male rabbits after oral doses of 100 mg/kg. 2. Both piperonylidene acetone and piperonyl acetone were extensively excreted as 4-(3,4-methylenedioxyphenyl)-butan-2-ol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service