Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W290500

Sigma-Aldrich

Pine needle oil

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2905
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Kosher
natural
biological source:
Pinus spp.
food allergen:
no known allergens

biological source

Pinus spp.

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012

form

liquid

composition

d-Limonene, 3.5%

refractive index

n20/D 1.470

density

0.900 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; balsamic

General description

Pine needle oil may be composed of oils from different pine species such as Pinus elliottii, Pinus palustris, Pinus taeda, Pinus serotina and Pinus clausa. It is generally used as a fragrance ingredient in soaps and perfumes.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

105.0 °F - closed cup

Flash Point(C)

40.56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composition of pine needle oil.
Joye Jr NM, et al.
Journal of Chromatographic Science, 10(9), 590-592 (1972)
David William Green et al.
Scientific reports, 7, 41023-41023 (2017-01-26)
The external epithelial surfaces of plants and animals are frequently carpeted with small micro- and nanostructures, which broadens their adaptive capabilities in challenging physical habitats. Hairs and other shaped protuberances manage with excessive water, light contaminants, predators or parasites in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service