Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W282537

Sigma-Aldrich

Orange oil

cold-pressed, Brazil origin, FG

Synonym(s):

Citrus aurantium, Citrus sinensis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2825
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Citrus sinensis (L.) Osbeck

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.230
FDA 21 CFR 182.20

optical activity

[α]20/D +97°, neat

quality

cold-pressed

origin

Brazil origin

refractive index

n20/D 1.473

bp

175 °C

density

0.843 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orange

Preparation Note

Extraction method: cold pressed

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean J Pendleton et al.
Journal of food science, 77(6), M308-M311 (2012-05-16)
Plant essential oils have previously been shown to exhibit antimicrobial activities against various microorganisms. In this study, cold pressed terpeneless Valencia orange oil (CPTVO) was examined at various temperatures (37, 10, and 4 °C) to determine its antimicrobial activity against
David Julian McClements et al.
Journal of food science, 77(1), C33-C38 (2011-12-03)
Beverage emulsions containing flavor oils that have a relatively high water-solubility are unstable to droplet growth due to Ostwald ripening. The aim of this study was to improve the stability of model beverage emulsions to this kind of droplet growth
Mário Tanomaru-Filho et al.
Brazilian dental journal, 21(1), 46-49 (2010-05-14)
The aim of this study was to evaluate the effectiveness of 3 solvents (eucalyptol, orange oil, and xylol) on 2 types of gutta-percha (conventional and thermoplastic) and Resilon. Specimens (10 mm diameter x 1 mm thick; n=7 per condition) were
Jong-Hwan Lim et al.
Experimental animals, 61(1), 71-75 (2012-02-02)
This study was conducted to evaluate the oral absorption of enrofloxacin (ENFX) in rats when administered with orange oil or its main component, limonene. Compared with the group administered ENFX alone, the ENFX + limonene group did not show any
Kristina Kejlová et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(8), 2084-2089 (2010-08-07)
The aim of this study, linked-up with a previous study on bergamot oils, was the evaluation of phototoxic potential of essential oils (orange, lemon and Litsea cubeba), used as cosmetic ingredients. The applied tiered testing strategy included chemical analysis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service