Skip to Content
MilliporeSigma
All Photos(1)

Documents

W278300

Sigma-Aldrich

1,3-Nonanediol acetate, mixed esters

≥80%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH2CH(OR)CH2CH2OR, R = H or -COCH3
CAS Number:
Molecular Weight:
202.29
FEMA Number:
2783
EC Number:
Council of Europe no.:
2075
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.225

biological source

synthetic

Assay

≥80%

refractive index

n20/D 1.446 (lit.)

bp

265 °C (lit.)

density

0.959 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CCCCCCC(O)CCO.CCCCCCC(CCOC(C)=O)OC(C)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service