Skip to Content
MilliporeSigma
All Photos(1)

Documents

W268003

Sigma-Aldrich

2-Methylanisole

≥99%, FG

Synonym(s):

o-Methylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OCH3
CAS Number:
Molecular Weight:
122.16
FEMA Number:
2680
EC Number:
Council of Europe no.:
187
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.014
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥99%

refractive index

n20/D 1.516 (lit.)

bp

170-172 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; earthy; walnut

SMILES string

COc1ccccc1C

InChI

1S/C8H10O/c1-7-5-3-4-6-8(7)9-2/h3-6H,1-2H3

InChI key

DTFKRVXLBCAIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Methylanisole is found in mastic oils, virgin olive oils and frankincense.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frankincense and myrrh resources of Ethiopia: II. Medicinal and industrial uses.
Lemenith M & Teketay D.
SINET: Ethiopian Journal of Science, 26(2), 161-172 (2003)
Quality profile determination of Chios mastic gum essential oil and detection of adulteration in mastic oil products with the application of chiral and non-chiral GC?MS analysis.
Paraschos S, et al.
Fitoterapia, 114, 12-17 (2016)
Application of solid-phase microextraction to the analysis of volatile compounds in virgin olive oils from five new cultivars
Baccouri B, et al.
Food Chemistry, 102(3), 850-856 (2007)
Elodie Guyonnet Bilé et al.
ChemSusChem, 5(1), 91-101 (2012-01-18)
Optically active amphiphilic compounds derived from N-methylephedrine, N-methylprolinol, or cinchona derivatives possessing bromide or chiral lactate counterions were efficiently used as protective agents for rhodium(0) nanoparticles. The full characterization of these surfactants and the obtained nanocatalysts was performed by means
L Higgins et al.
Archives of biochemistry and biophysics, 385(1), 220-230 (2001-05-22)
Regioselectivity is used to determine the absolute energetic differences for four different reactions catalyzed by P450. Abstraction of a hydrogen from a benzylic carbon containing a chlorine has a 1.0 kcal/mol lower barrier than abstraction from a simple benzylic carbon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service