Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W263117

Sigma-Aldrich

Lime oil

expressed, Mexico or Tahiti origin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2631
MDL number:
UNSPSC Code:
12164502

biological source

Citrus aurantifolia

grade

Kosher

reg. compliance

FDA 21 CFR 172.230

optical activity

[α]20/D +37.5°, neat

origin

Mexico or Tahiti origin

refractive index

n20/D 1.474-1.477 (lit.)
n20/D 1.476 (lit.)

bp

176 °C

density

0.861 g/mL at 25 °C
0.876 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

lime

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: cold pressed

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

120.0 °F - closed cup

Flash Point(C)

48.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service