Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W259802

Sigma-Aldrich

Jasmin absolute

FG

Synonym(s):

Jasmine oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2598
MDL number:
UNSPSC Code:
12164502
grade:
FG
Halal
Kosher
biological source:
Jasminum grandiflorum L.
food allergen:
no known allergens

biological source

Jasminum grandiflorum L.

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

refractive index

n20/D 1.50 (lit.)

density

0.947 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

jasmine

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: solvent extraction

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

198.0 °F - closed cup

Flash Point(C)

92.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Lis-Balchin et al.
Phytotherapy research : PTR, 16(5), 437-439 (2002-08-31)
Jasmine (Jasminum grandiflorum L.) is used in aromatherapy as a holistic treatment for apathy, hysteria, uterine disorders and childbirth, muscle relaxation and coughs. Its stimulant nature, on inhalation, has been shown both in animals and man. Jasmine has a spasmolytic
Tapanee Hongratanaworakit
Natural product communications, 5(1), 157-162 (2010-02-27)
The aim of this study was to investigate the effect of aromatherapy massage with jasmine oil (Jasminum sambac L., Oleaceae) on humans. Human autonomic parameters, i.e. blood pressure, pulse rate, blood oxygen saturation, breathing rate, and skin temperature, were recorded
Alfreda Wei et al.
Journal of agricultural and food chemistry, 55(5), 1737-1742 (2007-02-14)
Thirteen essential oils were examined for their antioxidant activity using three different assay systems. Jasmine, parsley seed, rose, and ylang-ylang oils inhibited hexanal oxidation by over 95% after 40 days at a level of 500 microg/mL in the aldehyde/carboxylic acid
J Ilmberger et al.
Chemical senses, 26(3), 239-245 (2001-04-05)
Scientific research on the effects of essential oils on human behavior lags behind the promises made by popular aromatherapy. Nearly all aspects of human behavior are closely linked to processes of attention, the basic level being that of alertness, which
"Jasmine--the lactifuge".
S R Acharya
The Journal of the Association of Physicians of India, 35(7), 543-544 (1987-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service