Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W259200

Sigma-Aldrich

Immortelle absolute

natural

Synonym(s):

Helichrysum oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2592
EC Number:
MDL number:
UNSPSC Code:
12164502

biological source

Helichrysum angustifolium

grade

Kosher
natural

reg. compliance

FDA 21 CFR 182.20

refractive index

n20/D 1.479

density

0.894 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

spicy; woody

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3 - Resp. Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service