Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W256501

Sigma-Aldrich

Hexyl acetate

≥98%, FCC, FG

Synonym(s):

Capryl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

W256501-SAMPLE-K
$74.30
1 KG
$102.00
5 KG
$251.00
20 KG
$590.00

$74.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W256501-SAMPLE-K
$74.30
1 KG
$102.00
5 KG
$251.00
20 KG
$590.00

About This Item

Linear Formula:
CH3COO(CH2)5CH3
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2565
Beilstein:
1747138
EC Number:
Council of Europe no.:
196
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.006
NACRES:
NA.21
Organoleptic:
apple; green; fruity; pear; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

$74.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.409 (lit.)

bp

168-170 °C (lit.)

mp

−80 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; fruity; pear; sweet

SMILES string

CCCCCCOC(C)=O

InChI

1S/C8H16O2/c1-3-4-5-6-7-10-8(2)9/h3-7H2,1-2H3

InChI key

AOGQPLXWSUTHQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Bioprospecting of sourdough microbial species from artisan bakeries in the city of Valencia.: Investigating microbial diversity in sourdough, this study identifies hexyl acetate as a key volatile contributing to the unique flavors of artisan bread. The research underscores the importance of microbial bioprospecting in enhancing food flavors and textures (Sanmartín et al., 2024).

  • Bioactivities of scent gland chemicals from Mictis fuscipes Hsiao (Hemiptera: Coreidae) on Solenopsis invicta Buren (Hymenoptera: Formicidae).: This study details the defensive chemical strategies of Mictis fuscipes, focusing on hexyl acetate′s role in repelling predators and pests. Such insights are valuable for developing natural pest control methods and understanding ecological interactions (Zhong et al., 2024).

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Wu et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(12), 1855-1863 (2015-09-12)
The multispecific organic anion drug transporters OAT6 (SLC22A20) and OAT1 (SLC22A6) are expressed in nasal epithelial cells and both can bind odorants. Sequence analysis of OAT6 revealed an evolutionarily conserved 79-amino acid (AA) fragment present not only in OAT6 but
Zhilin Zhang et al.
Journal of insect science (Online), 14 (2014-12-21)
The morphological structure of the metathoracic scent glands (MTGs) in Adelphocoris suturalis was observed by utilizing scanning electron microscope (SEM). Also, the secretions of MTGs in male and female were analyzed by using gas chromatography-mass spectrometry (GC-MS). The result showed
Xiaobing Pang
Journal of environmental sciences (China), 32, 196-206 (2015-06-05)
Green leaf volatiles (GLVs) emitted by plants after stress or damage induction are a major part of biogenic volatile organic compounds (BVOCs). Proton transfer reaction time-of-flight mass spectrometry (PTR-TOF-MS) is a high-resolution and sensitive technique for in situ GLV analyses
Kye Chung Park et al.
Journal of insect physiology, 78, 33-46 (2015-05-06)
Phytophagous insects detect volatile compounds produced by host and non-host plants, using species-specific sets of olfactory receptor neurons (ORNs). To investigate the relationship between the range of host plants and the profile of ORNs, single sensillum recordings were carried out
Caroline Mongruel Eleutério Dos Santos et al.
Journal of food science, 80(6), C1170-C1177 (2015-04-30)
The amino acid profile in dessert apple must and its effect on the synthesis of fusel alcohols and esters in cider were established by instrumental analysis. The amino acid profile was performed in nine apple musts. Two apple musts with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service