Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W251607

Sigma-Aldrich

Geranyl phenylacetate

stabilized, FCC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
272.38
FEMA Number:
2516
EC Number:
Council of Europe no.:
231
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.704

biological source

synthetic

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

contains

alpha-tocopherol, synthetic as stabilizer

impurities

~25% citronellyl phenylacetate

refractive index

n20/D 1.509 (lit.)

bp

278 °C (lit.)

density

0.972 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

floral

SMILES string

C\C(C)=C\CC\C(C)=C\COC(=O)Cc1ccccc1

InChI

1S/C18H24O2/c1-15(2)8-7-9-16(3)12-13-20-18(19)14-17-10-5-4-6-11-17/h4-6,8,10-12H,7,9,13-14H2,1-3H3/b16-12+

InChI key

UXAIJXIHZDZMSK-FOWTUZBSSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service