Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W245518

Sigma-Aldrich

Ethyl 3-phenylpropionate

≥98%, FG

Synonym(s):

Ethyl hydrocinnamate, Ethyl 3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2COOC2H5
CAS Number:
Molecular Weight:
178.23
FEMA Number:
2455
Beilstein:
1910432
EC Number:
Council of Europe no.:
429
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.747
NACRES:
NA.21
Organoleptic:
ethereal; fruity
grade:
FG
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.494 (lit.)

bp

247-248 °C (lit.)

density

1.01 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

ethereal; fruity

SMILES string

CCOC(=O)CCc1ccccc1

InChI

1S/C11H14O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3

InChI key

JAGZUIGGHGTFHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 3-phenylpropionate is a volatile flavor compound that is reported to occur in muskmelon and red fermented soybean curds.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile components in aqueous essence and fresh fruit of Cucumis melo cv. Athena (muskmelon) by GC-MS and GC-O.
Jordan MJ, et al.
Journal of Agricultural and Food Chemistry, 49(12), 5929-5933 (2001)
The FEMA GRAS assessment of cinnamyl derivatives used as flavor ingredients.
Adams TB, et al.
Food And Chemical Toxicology, 42(2), 157-185 (2004)
Enbo Xu et al.
Journal of food science, 80(7), C1476-C1489 (2015-06-20)
Enzymatic extrusion, instead of traditional steam cooking, to treat rice is an efficient and alternative pretreatment for Chinese rice wine fermentation. In order to determine the formation of volatiles in enzymatic extrusion-processed rice wine (EE), and to confirm its characteristic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service