Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W244601

Sigma-Aldrich

Ethyl nitrite solution

10-20 wt. % in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5ONO
CAS Number:
Molecular Weight:
75.07
MDL number:
Flavis number:
16.017

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

concentration

10-20 wt. % in ethanol

refractive index

n20/D 1.360

density

0.792 g/mL at 25 °C

Organoleptic

nearly

SMILES string

CCON=O

InChI

1S/C2H5NO2/c1-2-5-3-4/h2H2,1H3

InChI key

QQZWEECEMNQSTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C MacDonald et al.
Acta chemica Scandinavica. Series B: Organic chemistry and biochemistry, 35(7), 485-487 (1981-01-01)
Gas chromatography--mass spectrometry demonstrates the presence of ethyl nitrite in the breath of volunteers subjects who smoke and drink concurrently. The mutagenic compound is quantitated at the ppb level by gas chromatography with the selective electron capture detector. The compound
Alessio Baccarani et al.
Journal of reconstructive microsurgery, 23(5), 257-262 (2007-09-19)
Vasospasm leading to thrombosis is a major cause of free flap failure. Flap blood flow may improve by application of vasodilating agents to the pedicle. The purpose of this study is to evaluate the effect of a novel S-nitrosylating agent
Martin P Moya et al.
Lancet (London, England), 360(9327), 141-143 (2002-07-20)
Inhaled nitric oxide is used to alleviate pulmonary hypertension and hypoxaemia, but generates toxic free radicals and oxides of nitrogen (NO(x)), which can cause rebound-hypoxia and additional pulmonary and other morbidity. To address these problems, we assessed the efficacy of
Basil M Yurcisin et al.
Annals of surgery, 257(5), 971-977 (2013-01-31)
To determine if reduction in nitric oxide bioactivity contributes to the physiological instability that occurs after brain death and, if so, to also determine in this setting whether administration of a renitrosylating agent could improve systemic physiological status. Organ function
Richard L Auten et al.
American journal of respiratory and critical care medicine, 176(3), 291-299 (2007-05-05)
Inhaled nitric oxide (NO) has been used to prevent bronchopulmonary dysplasia, but with variable results. Ethyl nitrite (ENO) forms S-nitrosothiols more readily than does NO, and resists higher-order nitrogen oxide formation. Because S-nitrosylation is a key pathway mediating many NO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service