Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W243108

Sigma-Aldrich

Ethyl cyclohexanepropionate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
FEMA Number:
2431
EC Number:
Council of Europe no.:
2095
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.488

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

Assay

≥99%

refractive index

n20/D 1.448 (lit.)

bp

91-94 °C/8 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

CCOC(=O)CCC1CCCCC1

InChI

1S/C11H20O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h10H,2-9H2,1H3

InChI key

NRVPMFHPHGBQLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service