Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W242500

Sigma-Aldrich

2-Ethylbutyl acetate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH(C2H5)2
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2425
EC Number:
Council of Europe no.:
215
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.025
NACRES:
NA.21

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.41 (lit.)

bp

160 °C/740 mmHg (lit.)

density

0.876 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; oily; ethereal; earthy; citrus; peach; sweet

SMILES string

CCC(CC)COC(C)=O

InChI

1S/C8H16O2/c1-4-8(5-2)6-10-7(3)9/h8H,4-6H2,1-3H3

InChI key

HQLKZWRSOHTERR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 10 ppm

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 27(4), 68-68 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service