Skip to Content
MilliporeSigma
All Photos(1)

Documents

W232106

Sigma-Aldrich

Clary sage oil

Synonym(s):

Salvia Sclarea

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2321
MDL number:
UNSPSC Code:
12164502

grade

Kosher

reg. compliance

FDA 21 CFR 182.60

optical activity

[α]20/D −10.5°, neat

origin

USA origin

refractive index

n20/D 1.458 (lit.)

bp

210 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; honey; woody; minty; citrus; fruity; spicy; floral; sweet

Biochem/physiol Actions

Taste at 20 ppm

Preparation Note

Extraction method: steam distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 30, 56-56 (2005)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 41-41 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service