Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W231614

Sigma-Aldrich

Citronellyl propionate

natural (US), ≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
212.33
FEMA Number:
2316
EC Number:
Council of Europe no.:
410c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.129

grade

FG
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.444 (lit.)

bp

242 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CCC(=O)OCCC(C)CC\C=C(\C)C

InChI

1S/C13H24O2/c1-5-13(14)15-10-9-12(4)8-6-7-11(2)3/h7,12H,5-6,8-10H2,1-4H3

InChI key

POPNTVRHTZDEBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service