Skip to Content
MilliporeSigma
All Photos(1)

Documents

W230804

Sigma-Aldrich

Citronella oil, Chinese 85/35

FG

Synonym(s):

Citronella grass oil, Cymbopogon winterianus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2308
MDL number:
UNSPSC Code:
12164502

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

refractive index

n20/D 1.485 (lit.)

bp

222 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

lemon

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Miró Specos et al.
Transactions of the Royal Society of Tropical Medicine and Hygiene, 104(10), 653-658 (2010-08-03)
Microcapsules containing citronella essential oil were prepared by complex coacervation and applied to cotton textiles in order to study the repellent efficacy of the obtained fabrics. Citronella released from treated textiles was indirectly monitored by the extractable content of its
Günter C Müller et al.
Journal of the American Mosquito Control Association, 24(1), 154-160 (2008-04-29)
The first goal of this study was to compare the degree of personal protection against biting insects provided by geraniol, linalool, and citronella candle (5%) vapors outdoors, where such products are commonly used. At a distance of 1.0 m, citronella
Kyu-Sik Chang et al.
Pest management science, 62(11), 1032-1038 (2006-08-09)
Patch and skin bioassays were used in laboratory and indoor tests to evaluate the repellency of (E)-cinnamaldehyde, identified in Cinnamomum cassia Blume bark and essential oil, and a cream containing 5% (w/w) cassia oil against Aedes aegypti (L.) females. Results
Sonali Sinha et al.
Journal of ethnopharmacology, 137(3), 1521-1527 (2011-09-08)
Essential oils of palmarosa and citronella have been extensively used in ancient Indian and South-east Asian traditional medicines. These essential oils have been reported to exhibit antimicrobial, anti parasitic effects against bacteria, yeasts, filamentous fungi, and viruses. In the present
Tracy M Katz et al.
Journal of the American Academy of Dermatology, 58(5), 865-871 (2008-02-15)
Arthropod bites remain a major cause of patient morbidity. These bites can cause local or systemic effects that may be infectious or inflammatory in nature. Arthropods, notably insects and arachnids, are vectors of potentially serious ailments including malaria, West Nile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service