Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W229202

Sigma-Aldrich

Cinnamon oil, ceylon type

FCC, FG

Synonym(s):

Cinnamomum zeylanicum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2292
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

grade

FG
Halal
Kosher

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

optical activity

[α]20/D −0.5°, neat

origin

Sri Lanka origin

refractive index

n20/D 1.533

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cinnamon; clove; spicy; woody

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service