Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W224111

Sigma-Aldrich

Cardamom oil

FCC, FG

Synonym(s):

Elletaria cardamomum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2241
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Elettaria cardamomum (L.)

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D +27°, neat

origin

Guatemala origin

refractive index

n20/D 1.464 (lit.)

bp

188 °C/760 mmHg (lit.)

density

0.924 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; spicy

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 5 ppm

Preparation Note

Extraction method: steam distillation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.0 °F - closed cup

Flash Point(C)

56.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 26, 60-60 (2001)
Mosciano, G.
Perfumer & Flavorist, 27, 92-92 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service