Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W221708

Sigma-Aldrich

Butyl valerate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CO2(CH2)3CH3
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2217
EC Number:
Council of Europe no.:
466
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.148
NACRES:
NA.21
Organoleptic:
green; fruity; tropical; pineapple; sweet
biological source:
synthetic
To inquire about this product W221708, please contact your local MilliporeSigma office or dealer. Contact Technical Service

biological source

synthetic

Quality Level

reg. compliance

FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.412 (lit.)

bp

186-187 °C (lit.)

density

0.868 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

green; fruity; tropical; pineapple; sweet

SMILES string

CCCCOC(=O)CCCC

InChI

1S/C9H18O2/c1-3-5-7-9(10)11-8-6-4-2/h3-8H2,1-2H3

InChI key

OKJADYKTJJGKDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service