Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W219010

Sigma-Aldrich

Butyl butyryllactate

natural (US), ≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(O2CCH2CH2CH3)CO2(CH2)3CH3
CAS Number:
Molecular Weight:
216.27
FEMA Number:
2190
EC Number:
Council of Europe no.:
2107c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.491
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural (US)

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.415-1.425 (lit.)

bp

90 °C/2 mmHg (lit.)

density

0.972 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

butter; cheese; creamy

SMILES string

CCCCOC(=O)C(C)OC(=O)CCC

InChI

1S/C11H20O4/c1-4-6-8-14-11(13)9(3)15-10(12)7-5-2/h9H,4-8H2,1-3H3

InChI key

NORZZKKLCYMBBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service