Skip to Content
MilliporeSigma
All Photos(1)

Documents

W216518

Sigma-Aldrich

(−)-Bornyl isovalerate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O2
CAS Number:
Molecular Weight:
238.37
FEMA Number:
2165
EC Number:
Council of Europe no.:
451
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.456

biological source

synthetic

Assay

≥97%

optical activity

[α]20/D −32.5°, neat

refractive index

n20/D 1.46 (lit.)

bp

255-260 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

camphoraceous

SMILES string

CC(C)CC(=O)OC1CC2CCC1(C)C2(C)C

InChI

1S/C15H26O2/c1-10(2)8-13(16)17-12-9-11-6-7-15(12,5)14(11,3)4/h10-12H,6-9H2,1-5H3

InChI key

MPYYVGIJHREDBO-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service