Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W215325

Sigma-Aldrich

Bergamot oil

Italy origin, rectified

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2153
MDL number:
UNSPSC Code:
12164502

biological source

Citrus aurantium L.

grade

Kosher

reg. compliance

FDA 21 CFR 182.20

origin

Italy origin

refractive index

n20/D 1.4665 (lit.)
n20/D 1.4675 (lit.)

bp

159 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orange

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Hee Hwang
Taehan Kanho Hakhoe chi, 36(7), 1123-1134 (2007-01-11)
The purpose of this study was to identify the effects of aromatherapy on blood pressure and stress responses of clients with essential hypertension. There were fifty-two subjects divided into an essential oil group, placebo group, and control group by random
Ji-Yeong Seo
Journal of Korean Academy of Nursing, 39(3), 357-365 (2009-07-03)
This study was done to examine the effects of aromatherapy on stress and stress responses in adolescents. A two-group cross-over design was used for this study. The experimental treatment was aroma essential oil inhalation and the placebo treatment was carrier
Giancarlo Fantin et al.
Journal of agricultural and food chemistry, 58(9), 5438-5443 (2010-04-16)
A new approach for removing monoterpenes (MTs) from bergamot oil by selective inclusion in deoxycholic acid (DCA) is proposed. The inclusion process is very efficient, the included fraction being composed mainly of limonene (71.7%) and gamma-terpinene (19.8%). On the other
Deborah H Ndao et al.
Psycho-oncology, 21(3), 247-254 (2012-03-03)
Though often lifesaving, stem cell transplantation (SCT) is a period of great distress for both child and parent. We conducted a double-blind, placebo-controlled randomized study evaluating the effect of the respiratory administration of bergamot essential oil on the anxiety, nausea
Emilia L Belsito et al.
Journal of agricultural and food chemistry, 55(19), 7847-7851 (2007-08-19)
The vacuum distillation of bergamot peels furnishes a high-quality essential oil that is totally bergapten-free. This oil was compared with that produced by distillation of cold-pressed oils and those commercially available. The oil obtained by vacuum distillation of the bergamot

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service