Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W215309

Sigma-Aldrich

Bergamot oil

Italy origin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2153
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

grade

Fragrance grade
Halal
Kosher

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

composition

Contains IFRA and EU 1223/2009 restricted Citral (Neral + Geranial) and EU1223/2009 restricted Linalool and Limonene (sum of D, L, and DL)

origin

Italy origin

refractive index

n20/D 1.4665 (lit.)
n20/D 1.4675 (lit.)

bp

159 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

limonene (sum of D, L and DL), linalool, citral (neral+geranial)

Organoleptic

orange; woody; citrus

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Fisher et al.
Letters in applied microbiology, 44(5), 495-499 (2007-04-25)
To test the effect of oils and vapours of lemon, sweet orange and bergamot and their components against three Arcobacter butzleri strains. The disc diffusion method was used to screen the oils and vapours against three strains of A. butzleri.
Jin-Hee Hwang
Taehan Kanho Hakhoe chi, 36(7), 1123-1134 (2007-01-11)
The purpose of this study was to identify the effects of aromatherapy on blood pressure and stress responses of clients with essential hypertension. There were fifty-two subjects divided into an essential oil group, placebo group, and control group by random
Tsukasa Sakurada et al.
Pharmacology, biochemistry, and behavior, 97(3), 436-443 (2010-10-12)
This study investigated the effect of bergamot essential oil (BEO) containing linalool and linalyl acetate as major volatile components in the capsaicin test. The intraplantar injection of capsaicin (1.6 μg) produced a short-lived licking/biting response toward the injected paw. The
Ji-Yeong Seo
Journal of Korean Academy of Nursing, 39(3), 357-365 (2009-07-03)
This study was done to examine the effects of aromatherapy on stress and stress responses in adolescents. A two-group cross-over design was used for this study. The experimental treatment was aroma essential oil inhalation and the placebo treatment was carrier
Deborah H Ndao et al.
Psycho-oncology, 21(3), 247-254 (2012-03-03)
Though often lifesaving, stem cell transplantation (SCT) is a period of great distress for both child and parent. We conducted a double-blind, placebo-controlled randomized study evaluating the effect of the respiratory administration of bergamot essential oil on the anxiety, nausea

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service