Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W215000

Sigma-Aldrich

Benzyl propionate

≥98%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2CH2C6H5
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2150
EC Number:
Council of Europe no.:
413
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.132
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.497 (lit.)

bp

222 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

benzyl alcohol

Organoleptic

banana; jasmine; floral; fruity; sweet

SMILES string

CCC(=O)OCc1ccccc1

InChI

1S/C10H12O2/c1-2-10(11)12-8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

VHOMAPWVLKRQAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service