Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W214115

Sigma-Aldrich

Benzyl isobutyrate

natural (US), ≥97%, FG

Synonym(s):

Benzyl-2-methyl propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O2
CAS Number:
Molecular Weight:
178.23
FEMA Number:
2141
EC Number:
Council of Europe no.:
301
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.426
NACRES:
NA.21
Organoleptic:
floral; fruity; sweet
grade:
FG
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Kosher
natural (US)

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.49 (lit.)

bp

238 °C (lit.)

density

1.003 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; fruity; sweet

SMILES string

CC(C)C(=O)OCc1ccccc1

InChI

1S/C11H14O2/c1-9(2)11(12)13-8-10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3

InChI key

UIKJRDSCEYGECG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service