Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W202002

Sigma-Aldrich

Allyl anthranilate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
FEMA Number:
2020
EC Number:
Council of Europe no.:
254
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.719

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

Assay

≥98%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.574 (lit.)

bp

105 °C/2 mmHg (lit.)

mp

224-228 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green

SMILES string

Nc1ccccc1C(=O)OCC=C

InChI

1S/C10H11NO2/c1-2-7-13-10(12)8-5-3-4-6-9(8)11/h2-6H,1,7,11H2

InChI key

UCANFCXAKYMFGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service