Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U2857

Sigma-Aldrich

Urethane

99%

Synonym(s):

Carbamic acid ethyl ester, Ethyl carbamate, Ethylurethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOC2H5
CAS Number:
Molecular Weight:
89.09
Beilstein:
635810
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor density

3.07 (vs air)

vapor pressure

10 mmHg ( 77.8 °C)

Assay

99%

bp

182-184 °C (lit.)

mp

48-50 °C (lit.)

SMILES string

CCOC(N)=O

InChI

1S/C3H7NO2/c1-2-6-3(4)5/h2H2,1H3,(H2,4,5)

InChI key

JOYRKODLDBILNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leon F Stankowski et al.
Mutagenesis, 30(3), 335-342 (2015-05-03)
As part of the international Pig-a validation trials, we examined the induction of Pig-a mutant reticulocytes and red blood cells (RET(CD59-) and RBC(CD59-), respectively) in peripheral blood of male Sprague Dawley(®) rats treated with urethane (25, 100 and 250mg/kg/day) or
Mehmet Arslan et al.
Environmental toxicology and pharmacology, 39(2), 787-793 (2015-03-04)
Deferasirox (commercially formulated as Exjade(®)) is one of the effective iron chelators used in treatment of iron overload diseases. In this study the effect of this substance for chromosome aberration, sister chromatid exchange and mitotic index was studied by in
Lourdes Santana et al.
Journal of medicinal chemistry, 51(21), 6740-6751 (2008-10-07)
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service