Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

THSD1445

ThalesNano MidiCart catalyst cartridge system

L × diam. 116 mm × 9.2 mm, 1%Pt/SiO2/polyethyleneimine

Synonym(s):

Heterogeneous catalysis system, safe Heterogeneous catalysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103817
NACRES:
NB.51

packaging

pack of 1 ea

manufacturer/tradename

ThalesNano THS D1445

L × diam.

116 mm × 9.2 mm

Looking for similar products? Visit Product Comparison Guide

General description

MiDiCarts are suitable for use only with Thalesnano H-Cube MidiCart reactors.

Legal Information

MidiCart is a trademark of ThalesNano Nanotechnology Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service