Key Documents
THS02134
ThalesNano CatCart® catalyst cartridge system, 70 mm L
5% Pd/CaCO3, poisoned with lead
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
UNSPSC Code:
60104500
NACRES:
NB.51
packaging
pack of 1 ea
manufacturer/tradename
ThalesNano THS 02134
diam.
4 mm
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)